accessibility menu, dialog, popup

UserName

NH2NON(R)O(R)OH(S)HO(S)HO

CAS RN 147-94-4

IUPAC Name: 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: cytarabine ara-C cytarabine liposome injection cytarbel arabitin cytosar-U β-arabinosylcytosine citarabina [inn-spanish] cytarabinoside cytosine arabinofuranoside
Molecular Weight (g/mol): 243.22
Molecular Formula: C9H13N3O5
InChi Key: UHDGCWIWMRVCDJ-CCXZUQQUSA-N
SMILES: NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)