missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytarabine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 214°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 243.22 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cytarabine |
Safety and Handling
| RTECSNumber | HA5425000 |
| TSCA | No |