accessibility menu, dialog, popup

UserName

CAS RN 147-24-0

IUPAC Name: hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
Synonyms: diphenhydramine hydrochloride
Molecular Weight (g/mol): 291.82
Molecular Formula: C17H22ClNO
InChi Key: PCHPORCSPXIHLZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (4)
  • (3)
Grade 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)