missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine Hydrochloride 98.0+%, TCI America™

Catalog No. D0423500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0423500G 500 g
D042325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0423500G Supplier TCI America Supplier No. D0423500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number MFCD00012479
InChI Key PCHPORCSPXIHLZ-UHFFFAOYSA-N
Synonym diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
PubChem CID 8980
ChEBI CHEBI:4637
IUPAC Name hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
SMILES [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 170°C
Color White
Quantity 500 g
Formula Weight 291.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenhydramine Hydrochloride
EINECSNumber (9)-0484&(9)-0690
RTECSNumber KR7000000
TSCA No