accessibility menu, dialog, popup

UserName

H3CCH3H3CCH3OOHOO

CAS RN 146670-40-8

IUPAC Name: 4-[2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-dioxolan-2-yl]benzoic acid
Synonyms: 4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)-1,3-dioxolan-2-yl]benzoic acid
Molecular Weight (g/mol): 380.48
Molecular Formula: C24H28O4
InChi Key: ZZUKALQMHNSWTK-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=CC(=CC=C12)C1(OCCO1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results