accessibility menu, dialog, popup

UserName

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHDy

CAS RN 14637-88-8

IUPAC Name: dysprosium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: dysprosium(3+) triacetylacetonate
Molecular Weight (g/mol): 459.83
Molecular Formula: C15H21DyO6
InChi Key: VABPDCUJKLETFM-UHFFFAOYSA-N
SMILES: [Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results