accessibility menu, dialog, popup

UserName

ZnSSNCH3SSNH3C

CAS RN 14634-93-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 457.99
Molecular Formula: C18H20N2S4Zn
InChi Key: KMNUDJAXRXUZQS-UHFFFAOYSA-L
SMILES: [Zn++].CCN(C([S-])=S)C1=CC=CC=C1.CCN(C([S-])=S)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)