accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 1462-96-0

IUPAC Name: 1-ethylcyclopentan-1-ol
Synonyms: 1-ethylcyclopentan-1-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: LPCWIFPJLFCXRS-UHFFFAOYSA-N
SMILES: CCC1(O)CCCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)