accessibility menu, dialog, popup

UserName

NH2NOO

CAS RN 1462-86-8

IUPAC Name: 3-aminopyridine-2-carboxylate
Synonyms: 3-aminopyridine-2-carboxylate
Molecular Weight (g/mol): 137.12
Molecular Formula: C6H5N2O2
InChi Key: BOOMHTFCWOJWFO-UHFFFAOYSA-M
SMILES: NC1=CC=CN=C1C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)