CAS RN 1461-15-0
CAS RN 1461-15-0
IUPAC Name:
2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid
Synonyms:
calcein oftasceina oftasceine oftasceinum 奥荧光素
Molecular Weight (g/mol):
622.54
Molecular Formula:
C30H26N2O13
InChi Key:
DEGAKNSWVGKMLS-UHFFFAOYSA-N
SMILES:
OC(=O)CN(CC(O)=O)CC1=CC2=C(OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C22OC(=O)C3=CC=CC=C23)C=C1O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Fluorexon
CAS: 1461-15-0 Molecular Formula: C30H26N2O13 Molecular Weight (g/mol): 622.539 MDL Number: MFCD00005049 InChI Key: DEGAKNSWVGKMLS-UHFFFAOYSA-N Synonym: Bis[N, N-bis(carboxymethyl)aminomethyl]fluorescein,Calcein,Fluorescein Complexon PubChem CID: 65079 ChEBI: CHEBI:51903 IUPAC Name: 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O