accessibility menu, dialog, popup

UserName

CAS RN 14609-54-2

OHONHHOONHOONHNOHO

CAS RN 14609-54-2

IUPAC Name: 4-[7,12,17-tris(4-carboxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11(23),12,14,16(22),17,19-undecaen-2-yl]benzoic acid
Synonyms: 4-[7,12,17-tris(4-carboxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11(23),12,14,16(22),17,19-undecaen-2-yl]benzoic acid
Molecular Weight (g/mol): 790.79
Molecular Formula: C48H30N4O8
InChi Key: SMOZAZLNDSFWAB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results