accessibility menu, dialog, popup

UserName

PdClClNNH3CCH3

CAS RN 14592-56-4

IUPAC Name: bis(acetonitrile); dichloropalladiumbis(ylium)
Synonyms: bis(acetonitrile); dichloropalladiumbis(ylium)
Molecular Weight (g/mol): 259.42
Molecular Formula: C4H6Cl2N2Pd
InChi Key: XGPJSRXOBCGBMN-UHFFFAOYSA-L
SMILES: CC#N.CC#N.Cl[Pd++]Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)