accessibility menu, dialog, popup

UserName

CAS RN 1455-77-2

IUPAC Name: 1H-1,2,4-triazole-3,5-diamine
Synonyms: 1H-1,2,4-triazole-3,5-diamine
Molecular Weight (g/mol): 99.1
Molecular Formula: C2H5N5
InChi Key: PKWIYNIDEDLDCJ-UHFFFAOYSA-N
SMILES: NC1=NC(N)=NN1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)