accessibility menu, dialog, popup

UserName

CAS RN 14542-93-9

IUPAC Name: 2-isocyano-2,4,4-trimethylpentane
Synonyms: 2-isocyano-2,4,4-trimethylpentane
Molecular Weight (g/mol): 139.24
Molecular Formula: C9H17N
InChi Key: YVPXQMYCTGCWBE-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)[N+]#[C-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results