CAS RN 144875-48-9
CAS RN 144875-48-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Resiquimod 98.0+%, TCI America™
CAS: 144875-48-9 Molecular Formula: C17H22N4O2 Molecular Weight (g/mol): 314.389 MDL Number: MFCD00937759 InChI Key: BXNMTOQRYBFHNZ-UHFFFAOYSA-N Synonym: resiquimod,1-4-amino-2-ethoxymethyl-1h-imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol,resiquimod inn,unii-v3dmu7pvxf,v3dmu7pvxf,resiquimod r-848,4-amino-2-ethoxymethyl-a,a-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol,alpha-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol,1-4-amino-2-ethoxymethyl imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol PubChem CID: 159603 ChEBI: CHEBI:36706 IUPAC Name: 1-[4-amino-2-(ethoxymethyl)imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol SMILES: CCOCC1=NC2=C(N1CC(C)(C)O)C3=CC=CC=C3N=C2N