CAS RN 14476-37-0
IUPAC Name:
undecan-4-one
Synonyms:
undecan-4-one
Molecular Weight (g/mol):
170.3
Molecular Formula:
C11H22O
InChi Key:
NBSLHMOSERBUOV-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)CCC
1
–
1
of
1
results
1
4-Undecanone 95.0+%, TCI America™
CAS: 14476-37-0 Molecular Formula: C11H22O Molecular Weight (g/mol): 170.30 MDL Number: MFCD00048923 InChI Key: NBSLHMOSERBUOV-UHFFFAOYSA-N Synonym: 4-undecanone,heptyl propyl ketone,n-heptyl n-propyl ketone,acmc-1c992 PubChem CID: 84467 IUPAC Name: undecan-4-one SMILES: CCCCCCCC(=O)CCC