CAS RN 1445-78-9
IUPAC Name:
2,5-diphenylthiophene
Synonyms:
2,5-diphenylthiophene
Molecular Weight (g/mol):
236.33
Molecular Formula:
C16H12S
InChi Key:
HWKZNRABKQDKTC-UHFFFAOYSA-N
SMILES:
S1C(=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,5-Diphenylthiophene 98.0+%, TCI America™
CAS: 1445-78-9 Molecular Formula: C16H12S Molecular Weight (g/mol): 236.332 InChI Key: HWKZNRABKQDKTC-UHFFFAOYSA-N PubChem CID: 74050 IUPAC Name: 2,5-diphenylthiophene SMILES: C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=CC=C3