accessibility menu, dialog, popup

UserName

CAS RN 1445-75-6

IUPAC Name: bis(propan-2-yl) methylphosphonate
Synonyms: isopropyl methylphosphonate
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H17O3P
InChi Key: WOAFDHWYKSOANX-UHFFFAOYSA-N
SMILES: CC(C)OP(C)(=O)OC(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results