accessibility menu, dialog, popup

UserName

CAS RN 1445-39-2

IUPAC Name: 5-iodopyrimidin-2-amine
Synonyms: 5-iodopyrimidin-2-amine
Molecular Weight (g/mol): 221
Molecular Formula: C4H4IN3
InChi Key: HAFKCGZQRIIADX-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results