accessibility menu, dialog, popup

UserName

CAS RN 144173-85-3

IUPAC Name: 5-(tributylstannyl)pyrimidine
Synonyms: 5-(tributylstannyl)pyrimidine
Molecular Weight (g/mol): 369.14
Molecular Formula: C16H30N2Sn
InChi Key: QRDQHTJNKPXXRQ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)