accessibility menu, dialog, popup

UserName

CH3H3CH3CCH3

CAS RN 14411-56-4

IUPAC Name: 1-tert-butyl-3-ethylbenzene
Synonyms: 1-tert-butyl-3-ethylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: MUJPTTGNHRHIPH-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(=C1)C(C)(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)