missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-3-ethylbenzene, 98%

Catalog No. AAB2032514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2032514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2032514 Supplier Thermo Scientific Chemicals Supplier No. B2032514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14411-56-4 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14411-56-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00060845
InChI Key MUJPTTGNHRHIPH-UHFFFAOYSA-N
PubChem CID 139753
IUPAC Name 1-tert-butyl-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)C(C)(C)C

Specifications

Density 0.86
Boiling Point 205°C
Odor Mild
Quantity 25 g
Synonym 1-t-butyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene, benzene, 1-1,1-dimethylethyl-3-ethyl, benzene,1-1,1-dimethylethyl-3-ethyl, butylethylbenzene, m-tert-butyl-ethylbenzene, acmc-1cb13, 1-tert-butyl-3-ethyl-benzene, 1-1,1-dimethylethyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene 5g
Formula Weight 162.27
Percent Purity 98%
Chemical Name or Material 1-tert-Butyl-3-ethylbenzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only