missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-3-ethylbenzene 98.0+%, TCI America™

Catalog No. B17395ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17395ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17395ML Supplier TCI America Supplier No. B17395ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14411-56-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00060845
InChI Key MUJPTTGNHRHIPH-UHFFFAOYSA-N
Synonym 1-t-butyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene, benzene, 1-1,1-dimethylethyl-3-ethyl, benzene,1-1,1-dimethylethyl-3-ethyl, butylethylbenzene, m-tert-butyl-ethylbenzene, acmc-1cb13, 1-tert-butyl-3-ethyl-benzene, 1-1,1-dimethylethyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene 5g
PubChem CID 139753
IUPAC Name 1-tert-butyl-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)C(C)(C)C

Specifications

Color Colorless
Boiling Point 205°C
Quantity 5 mL
UN Number 3295
Formula Weight 162.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-tert-Butyl-3-ethylbenzene