accessibility menu, dialog, popup

UserName

HSBr

CAS RN 143888-85-1

IUPAC Name: (2-bromophenyl)methanethiol
Synonyms: (2-bromophenyl)methanethiol
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: UJNSDLRPHRMVGZ-UHFFFAOYSA-N
SMILES: SCC1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)