missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl mercaptan, 99%

Catalog No. AAH26247MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26247MD 250 mg
AAH2624703 1 g
AAH2624706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH26247MD Supplier Thermo Scientific Chemicals Supplier No. H26247MD

CAS: 143888-85-1 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143888-85-1
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD04038925
InChI Key UJNSDLRPHRMVGZ-UHFFFAOYSA-N
Synonym 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847
PubChem CID 2773673
IUPAC Name (2-bromophenyl)methanethiol
SMILES SCC1=CC=CC=C1Br

Specifications

Density 1.527
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 250 mg
UN Number UN2810
Sensitivity Air Sensitive
Formula Weight 203.1
Percent Purity 99%
Chemical Name or Material 2-Bromobenzyl mercaptan
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only