missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl Mercaptan 98.0+%, TCI America™

Catalog No. B37475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37475G 5 g
B374725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37475G Supplier TCI America Supplier No. B37475G

Chemical Identifiers

CAS 143888-85-1
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD04038925
InChI Key UJNSDLRPHRMVGZ-UHFFFAOYSA-N
Synonym 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847
PubChem CID 2773673
IUPAC Name (2-bromophenyl)methanethiol
SMILES SCC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 2810
Formula Weight 203.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromobenzyl Mercaptan