accessibility menu, dialog, popup

UserName

N(S)OHNOHOOOOH3CCH3CH3

CAS RN 143824-78-6

IUPAC Name: (2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-[1-(tert-butoxycarbonyl)indol-3-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid|FMOC-TRP(BOC)-OH
Molecular Weight (g/mol): 526.59
Molecular Formula: C31H30N2O6
InChi Key: ADOHASQZJSJZBT-SANMLTNESA-N
SMILES: CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)