accessibility menu, dialog, popup

UserName

CAS RN 14363-15-6

IUPAC Name: zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
Synonyms: zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
Molecular Weight (g/mol): 281.61
Molecular Formula: C10H16O5Zn
InChi Key: BNHYOPPPENBGNL-SUKNRPLKSA-L
SMILES: O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)