accessibility menu, dialog, popup

UserName

CAS RN 14338-82-0

IUPAC Name: butyl 2-({[(2-butoxy-2-oxoethyl)sulfanyl]methyl}sulfanyl)acetate
Synonyms: butyl 2-({[(2-butoxy-2-oxoethyl)sulfanyl]methyl}sulfanyl)acetate
Molecular Weight (g/mol): 308.45
Molecular Formula: C13H24O4S2
InChi Key: QCWRDNFBMVSDEY-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CSCSCC(=O)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results