accessibility menu, dialog, popup

UserName

CAS RN 1431612-23-5

CH3ONHH3CHNONNNNNCH3CH3CH3H3C

CAS RN 1431612-23-5

IUPAC Name: N-[(6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl]-1-(propan-2-yl)-6-{6-[4-(propan-2-yl)piperazin-1-yl]pyridin-3-yl}-1H-indazole-4-carboxamide
Synonyms: 1-isopropyl-6-[6-(4-isopropylpiperazin-1-yl)pyridin-3-yl]-N-[(6-methyl-2-oxo-4-propyl-1H-pyridin-3-yl)methyl]indazole-4-carboxamide
Molecular Weight (g/mol): 569.75
Molecular Formula: C33H43N7O2
InChi Key: DPJNKUOXBZSZAI-UHFFFAOYSA-N
SMILES: CCCC1=C(CNC(=O)C2=C3C=NN(C(C)C)C3=CC(=C2)C2=CN=C(C=C2)N2CCN(CC2)C(C)C)C(=O)NC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results