missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Venetoclax, 95%

Catalog No. AC468231000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468231000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468231000 Supplier Thermo Scientific™ Supplier No. 468231000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Specifications

Chemical Name or Material Venetoclax
CAS 1431612-23-5
Quantity 100 mg
Molecular Formula C45 H50 Cl N7 O7 S
Formula Weight 868.45
Physical Form Solid
Product Type Reagent
Purity 95%