CAS RN 143112-49-6
IUPAC Name:
7-(3-aminopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms:
7-(3-aminopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol):
311.36
Molecular Formula:
C12H17N5O3S
InChi Key:
SZJGHUNJZRXXCA-UHFFFAOYNA-N
SMILES:
CN(C)S(=O)(=O)C1=CC=C(N2CCC(N)C2)C2=NON=C12
1
–
1
of
1
results
1
(R)-(-)-DBD-APy 98.0+%, TCI America™
CAS: 143112-49-6 Molecular Formula: C12H17N5O3S Molecular Weight (g/mol): 311.36 MDL Number: MFCD00191379 InChI Key: SZJGHUNJZRXXCA-MRVPVSSYSA-N Synonym: (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole PubChem CID: 15682838 IUPAC Name: 7-[(3R)-3-aminopyrrolidin-1-yl]-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide SMILES: CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N3CCC(C3)N