accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 143-27-1

IUPAC Name: hexadecan-1-amine
Synonyms: palmitamine
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: FJLUATLTXUNBOT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCN
Boiling Point 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)