accessibility menu, dialog, popup

UserName

NiClClPPPhPhPhPhPhPh

CAS RN 14264-16-5

IUPAC Name: dichloronickelbis(ylium); bis(triphenylphosphane)
Synonyms: dichloronickelbis(ylium); bis(triphenylphosphine)
Molecular Weight (g/mol): 654.18
Molecular Formula: C36H30Cl2NiP2
InChi Key: VHQPRJDILRYZBG-UHFFFAOYSA-L
SMILES: Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)