accessibility menu, dialog, popup

UserName

CAS RN 1421-89-2

IUPAC Name: 2-(dimethylamino)ethyl acetate
Synonyms: 2-(dimethylamino)ethyl acetate
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: GOLSFPMYASLXJC-UHFFFAOYSA-N
SMILES: CN(C)CCOC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)