accessibility menu, dialog, popup

UserName

NiCH3CH3H3CHOH3CH3CCH3OOCH3H3CCH3HOH3CH3CCH3OO

CAS RN 1421-49-4

IUPAC Name: nickel(2+) bis(3,5-di-tert-butyl-4-hydroxybenzoate)
Synonyms: nickel(2+) bis(3,5-di-tert-butyl-4-hydroxybenzoate)
Molecular Weight (g/mol): 557.35
Molecular Formula: C30H42NiO6
InChi Key: VIUVLVWUWLHYGT-UHFFFAOYSA-L
SMILES: [Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)