missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%

Catalog No. AAA1688018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1688018 50 g
AAA1688009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1688018 Supplier Thermo Scientific Chemicals Supplier No. A1688018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1421-49-4 | C30H42NiO6 | 557.35 g/mol

3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1421-49-4
Molecular Formula C30H42NiO6
Molecular Weight (g/mol) 557.35
MDL Number MFCD00008827
InChI Key VIUVLVWUWLHYGT-UHFFFAOYSA-L
Synonym 3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681
PubChem CID 15007
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzoic acid
SMILES [Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O

Specifications

Melting Point 206°C to 209°C
Quantity 50 g
Beilstein 2216948
Solubility Information Insoluble in water.
Formula Weight 250.34
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 215-823-2
RTECSNumber DG6320000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only