missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. D238125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238125G 25 g
D2381250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D238125G Supplier TCI America Supplier No. D238125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1421-49-4
Molecular Formula C30H42NiO6
Molecular Weight (g/mol) 557.35
MDL Number MFCD00008827
InChI Key VIUVLVWUWLHYGT-UHFFFAOYSA-L
Synonym 3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681
PubChem CID 15007
IUPAC Name nickel(2+) bis(3,5-di-tert-butyl-4-hydroxybenzoate)
SMILES [Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 25 g
Formula Weight 250.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzoic Acid
RTECSNumber DG6320000
TSCA Yes