accessibility menu, dialog, popup

UserName

CAS RN 142-47-2

IUPAC Name: sodium 2-amino-4-carboxybutanoate
Synonyms: monosodium glutamate
Molecular Weight (g/mol): 169.11
Molecular Formula: C5H8NNaO4
InChi Key: LPUQAYUQRXPFSQ-UHFFFAOYNA-M
SMILES: [Na+].NC(CCC(O)=O)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)