accessibility menu, dialog, popup

UserName

NiOMoOOO

CAS RN 14177-55-0

IUPAC Name: nickel(2+) molybdenumbis(ylium)tetrakis(olate)
Synonyms: nickel(2+) molybdenumbis(ylium)tetrakis(olate)
Molecular Weight (g/mol): 218.64
Molecular Formula: MoNiO4
InChi Key: AXSAYZUZQSGYFU-UHFFFAOYSA-N
SMILES: [Ni++].[O-][Mo++]([O-])([O-])[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results