accessibility menu, dialog, popup

UserName

CAS RN 14165-55-0

IUPAC Name: tetrakis(ethanol) germanium
Synonyms: tetrakis(ethyl alcohol) germanium
Molecular Weight (g/mol): 256.91
Molecular Formula: C8H24GeO4
InChi Key: HLNJWGHNPKWEIJ-UHFFFAOYSA-N
SMILES: [Ge].CCO.CCO.CCO.CCO
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)