accessibility menu, dialog, popup

UserName

CAS RN 14150-95-9

OHNHN

CAS RN 14150-95-9

IUPAC Name: 2-imino-1,2-dihydropyridin-1-ol
Synonyms: 2-iminopyridin-1-ol
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: KTPMVZCGIJJWCD-UHFFFAOYSA-N
SMILES: ON1C=CC=CC1=N
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)