accessibility menu, dialog, popup

UserName

H3C(R)HOOHO(Z)

CAS RN 141-22-0

IUPAC Name: (9Z,12R)-12-hydroxyoctadec-9-enoic acid
Synonyms: ricinoleic acid l'acide ricinoleique acide ricinoleique kyselina ricinolova ricinolic acid 12-hydroxyoleic acid (r-(e))-isomer ricinoleic acid ricinoleic acid preparation ricinolsaeure ricinic acid
Molecular Weight (g/mol): 298.47
Molecular Formula: C18H34O3
InChi Key: WBHHMMIMDMUBKC-QJWNTBNXSA-N
SMILES: CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results