missing translation for 'onlineSavingsMsg'
Learn More

Ricinoleic acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013069
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013069 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013069 Supplier Merck Emd Millipore Supplier No. 90969100MG

Specifications

CAS 141-22-0
Density 0.940 g/mL (at 20°C)
Flash Point 224°C
Shipping Condition Wet Ice
Physical Form Neat
Linear Formula CH3(CH2)5 CH(OH)CH2 CH=CH(CH2)7 COOH
MDL Number MFCD00084840
Synonym (R)-12-Hydroxy-cis-9-octadecenoic acid; 12-Hydroxyoleic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 298.46
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less