missing translation for 'onlineSavingsMsg'
Learn More

Ricinoleic Acid 80.0+%, TCI America™

Catalog No. R0027500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0027500ML 500 mL
R002725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0027500ML Supplier TCI America Supplier No. R0027500ML

Chemical Identifiers

CAS 141-22-0
Molecular Formula C18H34O3
Molecular Weight (g/mol) 298.47
MDL Number MFCD00084840
InChI Key WBHHMMIMDMUBKC-QJWNTBNXSA-N
Synonym ricinoleic acid, ricinolic acid, ricinic acid, r,z-12-hydroxyoctadec-9-enoic acid, kyselina ricinolova, castor oil acid, 12-hydroxy-cis-9-octadecenoic acid, nouracid cs 80, 9-octadecenoic acid, 12-hydroxy-, r-z, kyselina 12-hydroxy-9-oktadecenova
PubChem CID 643684
ChEBI CHEBI:28592
IUPAC Name (9Z,12R)-12-hydroxyoctadec-9-enoic acid
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

Specifications

Quantity 500 mL
Formula Weight 298.47
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Ricinoleic Acid
EINECSNumber (2)-1340
RTECSNumber VJ3150000
TSCA Yes
Recommended Storage Refrigerator