accessibility menu, dialog, popup

UserName

CAS RN 14070-51-0

IUPAC Name: 2-chloro-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: 2-chloro-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Weight (g/mol): 217.62
Molecular Formula: C7H4ClNO3S
InChi Key: VKWMGUNWDFIWNW-UHFFFAOYSA-N
SMILES: ClN1C(=O)C2=CC=CC=C2S1(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results