accessibility menu, dialog, popup

UserName

H2NNH2OHNHNOHONHONH2NHONHONHOHNNH2ONNH2HNOHSOHOO(Z)

CAS RN 1405-37-4

IUPAC Name: 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
Synonyms: 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
Molecular Weight (g/mol): 766.79
Molecular Formula: C25H46N14O12S
InChi Key: AJQVUIHGEOLMDY-SOCRLDLMNA-N
SMILES: OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results