missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capreomycin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Capreomycin sulfate is a disulfate salt of the polypeptide antibiotic drug, capreomycin, that is administered either parenterally or in the form of aerosols. It is generally used against multidrug-resistant tuberculosis (MDR-TB).Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 1405-37-4 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Synonym | Capreomycin sulfate salt |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Grade | Certified Reference Material |