accessibility menu, dialog, popup

UserName

CH3CH3CH3OOH2C

CAS RN 14036-55-6

IUPAC Name: tert-butyl but-3-enoate
Synonyms: tert-butyl but-3-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: NGASWKRTXGWPNN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CC=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results