accessibility menu, dialog, popup

UserName

CAS RN 1400-62-0

IUPAC Name: 4,12-bis[(2,4-dihydroxy-6-methylphenyl)amino]-3,13-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1,3,6,9,12-pentaene-5,11-dione
Synonyms: 4,12-bis[(2,4-dihydroxy-6-methylphenyl)amino]-3,13-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1,3,6,9,12-pentaene-5,11-dione
Molecular Weight (g/mol): 500.51
Molecular Formula: C28H24N2O7
InChi Key: VPEASJIRGSVXBF-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(O)=C1NC1=C(C)C2=C3C(OC2=CC1=O)=CC(=O)C(NC1=C(C)C=C(O)C=C1O)=C3C
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Color 
  • (3)
Grade 
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)